Skip to Content
Merck
All Photos(5)

Key Documents

HPA009976

Sigma-Aldrich

Anti-ANGPTL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ANG-3, Anti-Angiopoietin-3, Anti-Angiopoietin-like 1, Anti-Angiopoietin-related protein 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LKMATRYRELEVKYASLTDLVNNQSVMITLLEEQCLRIFSRQDTHVSPPLVQVVPQHIPNSQQYTPGLLGGNEIQRDPGYPRDLMPPPDLATSPTKSPFKIPPVTFINEGPFKDCQQAKEAGHSVSGIYMIKPENSNGPMQLWCENS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANGPTL1(9068)

General description

ANGPTL1 (angiopoietin like 1), also called angiopoietin-3 (ANG3) is a member of the ANG family. It is composed of 491 amino acids, and is composed of the characteristic conserved coiled-coil domain and fibrinogen-like domain found in ANG family members, in its N- and C-terminals. It also contains the highly hydrophobic leader peptide in its N-terminal. It is a secretory protein and its mRNA is predominant in thyroid, heart, adrenal, placenta and small intestine in adult human tissues. It is also highly expressed in brain.

Immunogen

Angiopoietin-related protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ANGPTL1 (angiopoietin like 1) is a potent angiogenesis regulator. Its expression is negatively related with lymph node metastasis, invasion and poor prognosis. It acts as a tumor suppressor in breast and lung cancer cells. It suppresses SLUG (Snail homolog)-dependent EMT (epithelial mesenchymal transition) in lung cancer, thus, preventing cell motility. It is also thought to function as a transcription repressor. It forms heterodimers with Ear2 (V-erbA-related protein 2), and they may constitute a distinct orphan receptor signaling. It also functions as an agonist of Tie2, which is an endothelial receptor tyrosine kinase.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86560

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Avram et al.
The Journal of biological chemistry, 274(20), 14331-14336 (1999-05-13)
Members of the chicken ovalbumin upstream promoter-transcription factor (COUP-TF) subfamily of orphan nuclear receptors, which minimally includes COUP-TFI and ARP1, are highly expressed in brain and are generally considered to be constitutive repressors of transcription. We have used a yeast
Tsang-Chih Kuo et al.
The Journal of clinical investigation, 123(3), 1082-1095 (2013-02-26)
Angiopoietin-like protein 1 (ANGPTL1) is a potent regulator of angiogenesis. Growing evidence suggests that ANGPTL family proteins not only target endothelial cells but also affect tumor cell behavior. In a screen of 102 patients with lung cancer, we found that
I Kim et al.
FEBS letters, 443(3), 353-356 (1999-02-20)
Using homology-based PCR, we have isolated cDNA encoding a novel member (491 amino acids) of the angiopoietin (Ang) family from human adult heart cDNA and have designated it angiopoietin-3 (Ang3). The NH2-terminal and COOH-terminal portions of Ang-3 contain the characteristic
Hyuek Jong Lee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 18(11), 1200-1208 (2004-07-31)
The angiopoietin (Ang) family of growth factors includes Ang1, Ang2, Ang3, and Ang4, all of which bind to the endothelial receptor tyrosine kinase Tie2. Ang3 (mouse) and Ang4 (human) are interspecies orthologs. In experiments with human endothelial cell lines, Ang3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service