Skip to Content
Merck
All Photos(5)

Key Documents

HPA008741

Sigma-Aldrich

Anti-AGFG1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Arf-GAP domain and FG repeats-containing protein 1, Anti-HIV-1 Rev-binding protein, Anti-Nucleoporin-like protein RIP, Anti-Rev-interacting protein, Anti-Rev/Rex activation domain-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PQSTATANFANFAHFNSHAAQNSANADFANFDAFGQSSGSSNFGGFPTASHSPFQPQTTGGSAASVNANFAHFDNFPKSSSADFGTFNTSQSHQTASAVSKVSTNKAGLQTADKYAALANLDNIFSAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGFG1(3267)

General description

AGFG1 (ArfGAP with FG repeats 1) gene is localized to human chromosome 2q36.3 and is highly expressed in multiple human tissues. It interacts with Rev protein of human immunodeficiency virus (HIV), at its activation domain.

Immunogen

Arf-GAP domain and FG repeats-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AGFG1 (ArfGAP with FG repeats 1) is essential for the nuclear transport of HIV (human immunodeficiency virus) Rev-directed RNAs. It is also involved in the genome transport of influenza A virus from host nucleus to cell membrane. Polymorphisms in this gene are linked with the severity of airway hyperresponsiveness (AHR), which is a major feature of asthma. This protein is involved in endocytosis, where it acts as a partner of the tyrosine kinase substrate EPS15 (epidermal growth factor receptor pathway substrate 15). During spermatogenesis, this protein mediates fusion of preacrosomal vesicle required for the formation of acrosome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70274

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blanca E Himes et al.
BMC medical genetics, 14, 86-86 (2013-08-30)
Airway hyperresponsiveness (AHR), a primary characteristic of asthma, involves increased airway smooth muscle contractility in response to certain exposures. We sought to determine whether common genetic variants were associated with AHR severity. A genome-wide association study (GWAS) of AHR, quantified
Mathilde Chaineau et al.
The Journal of biological chemistry, 283(49), 34365-34373 (2008-09-30)
Human immunodeficiency virus Rev-binding protein (HRB), also called human Rev-interacting protein (hRIP) or Rev/Rex activation domain binding (RAB) is a partner of the tyrosine kinase substrate EPS15, and it has been recovered in the AP-2 interactome. EPS15 and AP-2 are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service