Skip to Content
Merck
All Photos(2)

Key Documents

HPA008719

Sigma-Aldrich

Anti-PLAU antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-U-plasminogen activator, Anti-Urokinase-type plasminogen activator precursor, Anti-uPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€479.00

€479.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€479.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€479.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

YLGRSRLNSNTQGEMKFEVENLILHKDYSADTLAHHNDIALLKIRSKEGRCAQPSRTIQTICLPSMYNDPQFGTSCEITGFGKENSTDYLYPEQLKMTVVKLISHRECQQPHYYGSEVTTKMLCAADPQWKTDSCQGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAU(5328)

General description

PLAU (plasminogen activator, urokinase) is a plasminogen activator, which is produced by renal and other cells. It is a serine protease, and acts as a ligand for UPAR (urokinase-type plasminogen receptor). This protein has a wide range of tissue expression.

Immunogen

Urokinase-type plasminogen activator precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PLAU (plasminogen activator, urokinase), post-activation, cleaves plasminogen to produce plasmin, which then degrades multiple constituents of the ECM (extracellular matrix). Along with UPAR and protease inhibitor PAI, it makes the UPA system, which is essential for controlled focal proteolysis and cell migration and adhesion. Therefore, it is also implicated in tumorigenesis, and is linked to gastric cancer (GC). The expression level of this system is also linked to the peritoneal metastasis of GC. In idiopathic membranous nephropathy, variants of this gene are linked with mortality and ESRD (end stage renal disease). Over-expression of this gene is linked to poor prognosis of pancreatic ductal adenocarcinoma (PDAC). It facilitates the stemness of cancer cells by interacting with LIM homeobox-2 (Lhx2), homeobox transcription factor A5 (HOXA5) and Hey proteins.[1] Studies in Northern Chinese Han population suggest SNPs (single nucleotide polymorphisms) in this gene are linked with the occurrence of oral tongue squamous cell carcinoma (OSCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71116

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youcheng Ding et al.
European journal of medical research, 18, 28-28 (2013-08-30)
It has been demonstrated that urokinase-type plasminogen activator (uPA) is involved in tumor cell metastasis by degrading the extracellular matrix. However, there is little direct evidence of clinical uPA system expression in peritoneal metastatic tissues of gastric cancer. The objective
Feng Zhong et al.
Asian Pacific journal of cancer prevention : APJCP, 14(2), 781-784 (2013-04-30)
The purpose of this study was to determine whether susceptibility to oral tongue squamous cell carcinoma (OSCC) is related to polymorphisms in the u-PA gene. We examined the rs2227564 C/T and rs2227562 G/A single nucleotide polymorphisms (SNPs) in 196 OSCC
Cheng-Hsu Chen et al.
BioMed research international, 2014, 425095-425095 (2014-05-14)
Idiopathic membranous nephropathy (MN) is one of the most common causes of nephrotic syndrome in adults, and 25% of MN patients proceed to ESRD. Urokinase plasminogen activator (uPA) may play an important role in reducing renal fibrosis. This study was
Swapna Asuthkar et al.
Molecular biology of the cell, 24(17), 2620-2632 (2013-07-19)
Pancreatic ductal adenocarcinoma (PDAC) is almost always lethal. One of the underlying reasons for this lethality is believed to be the presence of cancer stem cells (CSC), which impart chemoresistance and promote recurrence, but the mechanisms responsible are unclear. Recently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service