Skip to Content
Merck
All Photos(1)

Documents

HPA008194

Sigma-Aldrich

Anti-CLEC2B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Activation-induced C-type lectin antibody produced in rabbit, Anti-C-type lectin domain family 2 member B antibody produced in rabbit, Anti-C-type lectin superfamily member 2 antibody produced in rabbit, Anti-IFN-α2b-inducing-related protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LCPYDWIGFQNKCYYFSKEEGDWNSSKYNCSTQHADLTIIDNIEEMNFLRRYKCSSDHWIGLKMAKNRTGQWVDGATFTKSFGMRGSEGCAYLSDDGAATARCYTERKWICRKR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLEC2B(9976)

General description

C-type lectin domain family 2 member B (CLEC2B) is a leucocyte C-type lectin which is encoded by the NK gene complex. It is a 149-amino acid polypeptide with a short cytoplasmic domain having seven amino acids, a C-type lectin domain and a transmembrane region.

Immunogen

C-type lectin domain family 2 member B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

C-type lectin domain family 2 member B (CLEC2B) acts as a ligand for the activating NK receptor p80 (NKp80). Interleukin 2 (IL-2) and Interleukin 12 (IL-12) stimulate its expression on T cells. CLEC2B acts as an activation inducer molecule and an activation antigen.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70032.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Li et al.
European cytokine network, 11(4), 602-607 (2000-12-23)
IL-12 is a critical immunoregulatory cytokine that promotes cell-mediated immune responses by inducing the differentiation of Th1 cells. To better clarify the molecular basis of IL-12 action, we compared the gene expression in human T lymphocytes activated by IL-2 and
Differentially induced expression of C-type lectins in activated lymphocytes.
W Eichler at. al.
Journal of Cellular Biochemistry. Supplement, 81(S36), 201-208 (2001)
J Hamann et al.
Immunogenetics, 45(5), 295-300 (1997-01-01)
The NK gene complex on mouse chromosome 6 and its human homologue on chromosome 12 encode type II transmembrane proteins with a C-type lectin domain which trigger or inhibit target cell lysis by NK cells (NKR-P1, Ly49, NKG2, CD94) or
Jessica Spreu et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(11), 5100-5105 (2010-03-03)
Many well-known immune-related C-type lectin-like receptors (CTLRs) such as NKG2D, CD69, and the Ly49 receptors are encoded in the natural killer gene complex (NKC). Recently, we characterized the orphan NKC gene CLEC2A encoding for KACL, a further member of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service