Skip to Content
Merck
All Photos(3)

Key Documents

HPA008044

Sigma-Aldrich

Anti-SLC6A17 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Orphan sodium- and chloride-dependent neurotransmitter transporter NTT4, Anti-Solute carrier family 6 member 17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

MNEKCVVENAEKILGYLNTNVLSRDLIPPHVNFSHLTTKDYMEMYNVIMTVKEDQFSALGLDPCLLEDELDKSVQGTGLAFIAFTEAMTHFPASPFWSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Orphan sodium- and chloride-dependent neurotransmitter transporter NTT4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC6A17 (solute carrier family 6 (neutral amino acid transporter), member 17) gene belongs to a family of ′orphan transporters′ and functions as a sodium-dependent plasma membrane transporter. It facilitates the vesicular uptake of proline, glycine, leucine, and alanine and functions in synaptic transmission. It is localized to synaptic vesicles of glutamatergic and GABAergic neurons and function is specific transport of neurotransmitters. Mutations in this gene have been associated with autosomal-recessive intellectual disability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71191

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in SLC6A17 cause autosomal-recessive intellectual disability.
S Waltl
Clinical genetics, 88(2), 136-137 (2015-05-15)
Leonardo A Parra et al.
Molecular pharmacology, 74(6), 1521-1532 (2008-09-05)
Rxt1/NTT4 (SLC6A17) belongs to a gene family of "orphan transporters" whose substrates and consequently functions remain unidentified. Although Rxt1/NTT4 was previously thought to function as a sodium-dependent plasma membrane transporter, recent studies localized the protein to synaptic vesicles of glutamatergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service