Skip to Content
Merck
All Photos(2)

Key Documents

HPA007394

Sigma-Aldrich

Anti-NBL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-D1S1733E, Anti-DAN, Anti-DAND1, Anti-NB, Anti-NO3, Anti-neuroblastoma, suppression of tumorigenicity 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PPINKLALFPDKSAWCEAKNITQIVGHSGCEAKSIQNRACLGQCFSYSVPNTFPQSTESLVHCDSCMPAQSMWEIVTLECPGHEEVPRVDKLVEKILHCSCQACGKEPSHEGLSVYVQGEDGPGSQPGTHPHPHPHPHPGGQTPEPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NBL1(4681)

General description

NBL1 (neuroblastoma 1, DAN family BMP antagonist) gene is localized to human chromosome 1p36.13. The protein encoded by this gene is 180 amino acid (aa) long and belongs to the CAN (Cerberus and DAN) family of proteins.

Immunogen

neuroblastoma, suppression of tumorigenicity 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NBL1 (neuroblastoma 1, DAN family BMP antagonist) gene encodes a transcription factor that negatively regulates the cell cycle. It is found to be down-regulated in human colon cancer and may play a role in cancer development and progression. NBL1 is a secreted protein that serves as an antagonist against bone morphogenetic proteins (BMPs). It blocks the interaction of BMPs with their receptors. It is capable of stimulating differentiation of neuroblastoma cells in the presence of retinoic acid in vitro. It is also involved in osteoblast differentiation. This gene is observed to be over-expressed in pancreatic cancer and may serve as a potential marker for the diagnosis of pancreatic carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70260

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The NBL1 tumor suppressor gene is downregulated in colon cancer by promoter methylation.
Sung JC, et al.
Journal of the American College of Surgeons, 199, 92-92 (2004)
Neuroblastoma suppressor of tumorigenicity 1 is a circulating protein associated with progression to end-stage kidney disease in diabetes.
Kobayashi, et al.
Science Translational Medicine, 14, eabj2109-eabj2109 (2023)
Marek Olakowski et al.
Folia histochemica et cytobiologica, 47(2), 249-255 (2009-12-10)
The aim of the study was to analyze the gene expression profile of pancreatic cancer to derive novel molecular markers of this malignancy. The snap-frozen or RNA-later preserved samples of 18 pancreatic adenocarcinomas, 5 chronic pancreatitis cases and 6 specimens

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service