Skip to Content
Merck
All Photos(7)

Key Documents

HPA006757

Sigma-Aldrich

Anti-TSC22D4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TSC22 domain family protein 4 antibody produced in rabbit, Anti-TSC22-related-inducible leucine zipper protein 2 antibody produced in rabbit, Anti-Tsc-22-like protein THG-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TPLPSLRVEAEAGGSGARTPPLSRRKAVDMRLRMELGAPEEMGQVPPLDSRPSSPALYFTHDASLVHKSPDPFGAVAAQKFSLAHSMLAISGHLDSDDDSGSGSLVGIDNKIEQAMDLVKSHLMF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSC22D4(81628)

Related Categories

Immunogen

TSC22 domain family protein 4 recombinant protein epitope signature tag (PrEST)

Application

Anti-TSC22D4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

TSC22D4 (TSC22 domain family, member 4) is a heterodimeric leucine zipper-containing protein belonging to the TSC22 domain family. It contains a C-terminal TSC box region adjacent to the highly conserved leucine zipper domain. It has a unique quality of forming homodimer and heterodimer by binding to other leucine zipper-containing transcription factors. The tumor suppressor TSC22D4 binds to the heterologous DNA-binding domain to exert the transcriptional repressor activity. It catalyzes the 3′-5′ single guanine nucleotide addition to the 5′-end of nucleic acid substrates. It plays an imporatnt role in protein synthesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70971

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha J Hyde et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(47), 20305-20310 (2010-11-10)
All known DNA and RNA polymerases catalyze the formation of phosphodiester bonds in a 5' to 3' direction, suggesting this property is a fundamental feature of maintaining and dispersing genetic information. The tRNA(His) guanylyltransferase (Thg1) is a member of a
M T Fiorenza et al.
Gene, 278(1-2), 125-130 (2001-11-15)
Thg-1pit, a novel mouse gene, was detected in a screen for genes that are differentially expressed in the developing pituitary of wild-type and Lhx3 null mutant embryos. The predicted translation product of the Thg-1pit gene contains a C-terminal TSC-box adjacent
Cornelia Hömig-Hölzel et al.
The EMBO journal, 30(9), 1753-1765 (2011-03-31)
Oncogene-induced cellular senescence (OIS) is an increasingly recognized tumour suppressor mechanism that confines the outgrowth of neoplastic cells in vivo. It relies on a complex signalling network, but only few components have been identified so far. Gene-expression profiling revealed a
H A Kester et al.
The Journal of biological chemistry, 274(39), 27439-27447 (1999-09-17)
TGF-beta-stimulated clone-22 (TSC-22) encodes a leucine zipper-containing protein that is highly conserved during evolution. Two homologues are known that share a similar leucine zipper domain and another conserved domain (designated the TSC box). Only limited data are available on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service