Skip to Content
Merck
All Photos(4)

Documents

HPA006543

Sigma-Aldrich

Anti-CCT3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CCT-γ antibody produced in rabbit, Anti-T-complex protein 1 subunit gamma antibody produced in rabbit, Anti-TCP-1-γ antibody produced in rabbit, Anti-hTRiC5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

SLEYKKGESQTDIEITREEDFTRILQMEEEYIQQLCEDIIQLKPDVVITEKGISDLAQHYLMRANITAIRRVRKTDNNRIARACGARIVSRPEELREDDVGTGAGLLEIKKIGDEYFTFITDCKDPK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCT3(7203)

General description

CCT3 (chaperonin containing TCP1), a member of TCP-1 family of chaperonins, encodes a γ subunit of cytosolic chaperonin containing protein. This barrel-shaped molecule is widely distributed at the cytosol and centrosome of the human and mouse tissues.

Immunogen

T-complex protein 1 subunit gamma recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CCT3 (chaperonin containing TCP1) is mainly involved in the cellular protein folding homeostasis. It performs a pivotal role in the BBSome (Bardet-Biedl syndrome) assembly. In association with the BBS proteins, it mediates the transportation of ciliogenesis regulated vesicles to the cilia. Additionally, it is also associated with the p53-regulated several biological functions such as apoptosis, senescence, and cell cycle regulation. It is a potential biomarker for identifying the stage of hepatic tumor cholangiocarcinoma (CCA). Studies have suggested that CCT3 may regulate the activity of centrosome. It has clinic-pathological importance in the colon cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70722

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrated analyzes identify CCT3 as a modulator to shape immunosuppressive tumor microenvironment in lung adenocarcinoma.
Huang, et al.
BMC Cancer, 23, 241-241 (2023)
Yuan Shi et al.
Journal of gastrointestinal surgery : official journal of the Society for Surgery of the Alimentary Tract, 17(9), 1584-1591 (2013-07-23)
Cholangiocarcinoma (CCA) is becoming a common fatal hepatic tumor. Early detection of CCA is hampered by the absence of a sufficiently accurate and noninvasive diagnostic test. Proteomic analysis would be a powerful tool to identify potential biomarkers of this cancer.
Shao Chin Lee et al.
Cancer genomics & proteomics, 9(2), 101-108 (2012-03-09)
We recently reported that functional loss of p53 altered the responses of human HCT116 colon cancer cells to apoptosis triggers. To examine the molecular basis underlying the differential responses to drug treatment in the cancer cells, we performed a proteomic
N A Walkley et al.
The Biochemical journal, 313 ( Pt 2), 381-389 (1996-01-15)
We describe the cloning, DNA sequence analysis and mRNA expression analysis of human Cctg (HsCctg), a gene that encodes the gamma-subunit of the eukaryotic cytosolic 'chaperonin-containing TCP-1' (CCT). Partial clones representing the 3' region of HsCctg cDNA were isolated from
Seongjin Seo et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(4), 1488-1493 (2010-01-19)
Bardet-Biedl syndrome (BBS) is a human genetic disorder resulting in obesity, retinal degeneration, polydactyly, and nephropathy. Recent studies indicate that trafficking defects to the ciliary membrane are involved in this syndrome. Here, we show that a novel complex composed of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service