Skip to Content
Merck
All Photos(3)

Key Documents

HPA005544

Sigma-Aldrich

Anti-MTA1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Metastasis-associated protein MTA1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LEAVLRYLETHPRPPKPDPVKSVSSVLSSLTPAKVAPVINNGSPTILGKRSYEQHNGVDGNMKKRLLMPSRGLANHGQTRHMGPSRNLLLNGKSYPTKVRLIRGGSLPPVKRRRMNWIDAPDDVFYMATEETRKIRKLLSSSETK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTA1(9112)

General description

Metastasis tumor antigen 1 (MTA1) is a part of the nucleosome remodeling and deacetylating (NuRD) complex, and is also a candidate member for the metastasis-associated family of genes. MTA1 is localized to chromosomal position 14q32.3. This protein is localized in the nucleus. It has a molecular weight of ~82kDa and consists of 715 amino acids. The C-terminus of MTA1 consists of a proline-rich region or the SH3 (SRC homology 3)-binding domain. It is also composed of a leucine zipper, a putative DNA-binding zinc finger motif and five SPXX motifs.

Immunogen

Metastasis-associated protein MTA1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Metastasis tumor antigen 1 (MTA1) is a gene related to the progression of cancer. Its expression is up-regulated in various cancers, such as liver, breast, ovarian and colorectal cancer. As a component of nucleosome remodeling and deacetylating (NuRD) complex, it is thought to play a role in transcription control and chromatin remodeling. It acts as a repressor of estrogen receptor α (ERα), and an activator of Breast Cancer Amplified Sequence 3 (BCAS3). BCAS3 gene is overexpressed in breast cancers, and hence, MTA1 might play a role in tumorigenesis of breast cancer. MTA1 activates paired box gene 5 (Pax5), which in turn is up-regulated in B-cell lymphomas. Hence, MTA1 might be involved in lymphogenesis. When up-regulated in cells, this protein leads to chromosomal instability and aberrant number of centrosomes. This is achieved by the suppression of BRCA1, which is a tumor suppressor gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70514

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Jin Kang et al.
Cancer research, 74(5), 1484-1494 (2014-01-15)
Metastasis-associated protein 1 (MTA1) is a component of the nucleosome remodeling and histone deacetylase (HDAC) complex, which plays an important role in progression of breast cancer. Although MTA1 is known as a repressor of the transactivation function of estrogen receptor
Seetharaman Balasenthil et al.
Cancer research, 67(15), 7132-7138 (2007-08-03)
Previously, we have shown that metastasis-associated protein 1 (MTA1) overexpression in transgenic mice was accompanied by high incidence of spontaneous B-cell lymphomas including diffuse large B-cell lymphomas (DLBCL). To understand the molecular basis of lymphoma in MTA1-transgenic (MTA1-TG) mice, we
P R Molli et al.
Oncogene, 27(14), 1971-1980 (2007-10-09)
Metastasis-associated tumor antigen 1 (MTA1), a component of the nucleosome remodeling and deacetylating (NuRD) complex is routinely upregulated in several cancers. In the present study, we investigated the potential role of MTA1 in BRCA1 transcriptional repression and subsequent chromosomal instability.
Qingcui Song et al.
The international journal of biochemistry & cell biology, 45(7), 1439-1446 (2013-04-27)
Nasopharyngeal carcinoma (NPC) is prone to appearing regional lymph node and distant metastasis. And its underlying mechanism is unclear. Recent study suggests that overexpression of metastasis-associated gene 1 (MTA1) was independently associated with poorer distant metastasis-free survival in NPC. However
Anupama E Gururaj et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(17), 6670-6675 (2006-04-18)
Here we define a function of metastasis-associated protein 1 (MTA1), a presumed corepressor of estrogen receptor alpha (ERalpha), as a transcriptional activator of Breast Cancer Amplified Sequence 3 (BCAS3), a gene amplified and overexpressed in breast cancers. We identified BCAS3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service