Skip to Content
Merck
All Photos(5)

Key Documents

HPA003414

Sigma-Aldrich

Anti-PITPNM2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Membrane-associated phosphatidylinositol transfer protein 2, Anti-NIR-3, Anti-Phosphatidylinositol transfer protein, membrane-associated 2, Anti-Pyk2 N-terminal domain-interacting receptor 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PITPNM2(57605)

Immunogen

Membrane-associated phosphatidylinositol transfer protein 2 recombinant protein epitope signature tag (PrEST)

Sequence
PKDITKWSSNDLMDKIESPEPEDTQDGLYRQGAPEFRVASSVEQLNIIEDEVSQPLAAPPSKIHVLLLVLHGGTILDTGAGDPSSKKGDANTIANVFDTVMRVHYPSALGRLAIRLVPCPPVCSDAFALVSN

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PITPNM2 (Phosphatidylinositol transfer protein, membrane-associated 2) is localized on human chromosome 12p24. It is also known as NIR3, NIR-3, RDGB2 and RDGBA2. The protein binds to the amino-terminal domain of PYK2 through a conserved sequence motif located in the carboxy terminus. It consists of six putative transmembrane domains, a region homologous to phosphatidylinositol (PI) transfer protein, and an acidic domain. It is a calcium-binding protein that is involved in the transfer of phosphatidylinositol between membranes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86370

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Lev et al.
Molecular and cellular biology, 19(3), 2278-2288 (1999-02-18)
The protein tyrosine kinase PYK2 has been implicated in signaling pathways activated by G-protein-coupled receptors, intracellular calcium, and stress signals. Here we describe the molecular cloning and characterization of a novel family of PYK2-binding proteins designated Nirs (PYK2 N-terminal domain-interacting
L Ocaka et al.
Cytogenetic and genome research, 108(4), 293-302 (2005-01-04)
Eukaryotic proteins containing a phosphatidylinositol transfer (PITP) domain can be divided into two groups, one consisting of small soluble 35-kDa proteins and the other those that are membrane-associated and show sequence similarities to the Drosophila retinal degeneration B (rdgB) protein.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service