Skip to Content
Merck
All Photos(8)

Key Documents

HPA002238

Sigma-Aldrich

Anti-CCT5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

KIAA0098

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LDRGIHPIRIADGYEQAARVAIEHLDKISDSVLVDIKDTEPLIQTAKTTLGSKVVNSCHRQMAEIAVNAVLTVADMERRDVDFELIKVEGKVGGRLEDTKLIKGVIVDKDFSHPQM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCT5(22948)

Looking for similar products? Visit Product Comparison Guide

General description

CCT5 (chaperonin containing TCP1 subunit 5) is a cytosolic chaperonin containing protein involved in the organization of the cytoskeleton. It is localized in the microtubular structures and nucleus. It contains two superimposed rings, each with one cavity to hold other proteins during protein folding. It has a CCT 16-mer double-ring-like complex which differ CCT5 from others.

Immunogen

T-complex protein 1 subunit epsilon recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CCT5 (chaperonin containing TCP1 subunit 5) is a subunit of CCT. CCT is a molecular chaperone that helps in folding of newly synthesized proteins in eukaryotic cytosol such as actin, tubulin. The mRNA expression of CCT5 is up-regulated in p53-mutated breast tumors. Along with Bardet-Biedl syndrome (BBS) proteins, CCT5 facilitates BBSome assembly for transporting vesicles to the cilia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86551

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Himmelspach et al.
The Plant journal : for cell and molecular biology, 12(6), 1299-1310 (1998-02-05)
The cytosolic chaperonin containing TCP-1 (CCT) is known to keep fold cytoskeletal proteins and is involved in the proper organization of the cytoskeleton. These studies are based on the assumption that growth responses linked to structural rearrangement of the plant
José M Valpuesta et al.
FEBS letters, 529(1), 11-16 (2002-10-02)
Chaperonins are large oligomers made up of two superimposed rings, each enclosing a cavity used for the folding of other proteins. Among the chaperonins, the eukaryotic cytosolic chaperonin CCT is the most complex, not only with regard to its subunit
Hiroshi Kubota
Vitamins and hormones, 65, 313-331 (2002-12-17)
Molecular chaperones are a group of proteins that assists in the folding of newly synthesized proteins or in the refolding of denatured proteins. The cytosolic chaperonin-containing t-complex polypeptide 1 (CCT) is a molecular chaperone that plays an important role in
Asako Ooe et al.
Breast cancer research and treatment, 101(3), 305-315 (2006-07-06)
Present study was aimed to investigate the relationship of p53 mutation status with response to docetaxel in breast cancers. In addition, attempts were made to identify the genes differentially expressed between p53-wild and p53-mutated breast tumors and to study their
Seongjin Seo et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(4), 1488-1493 (2010-01-19)
Bardet-Biedl syndrome (BBS) is a human genetic disorder resulting in obesity, retinal degeneration, polydactyly, and nephropathy. Recent studies indicate that trafficking defects to the ciliary membrane are involved in this syndrome. Here, we show that a novel complex composed of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service