Skip to Content
Merck
All Photos(7)

Key Documents

HPA002191

Sigma-Aldrich

Anti-STX3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Syntaxin-3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KQLTQDDDTDAVEIAIDNTAFMDEFFSEIEETRLNIDKISEHVEEAKKLYSIILSAPIPEPKTKDDLEQLTTEIKKRANNVRNKLKSMEKHIEEDEVRSSADLRIRKSQHSVLSRKFVEVMTKYNEAQVDFRERSKGRIQRQLEITGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX3(6809)

General description

STX3 (syntaxin 3) belongs to the syntaxin family with seven isoforms, named as -1A, -1B, -2, -3, -4, -5, and -6. It is composed of a α-helical coiled-coil region and a highly hydrophobic region at the extreme carboxyl terminus. The α-helical region is responsible for protein-protein interactions whereas hydrophobic domain serves as a membrane anchor. Syntaxin 3 exists in two isoforms namely syntaxin 3A (previous syntaxin 3) and the novel syntaxin 3B.

Immunogen

Syntaxin-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In human STX3 (syntaxin 3), a plasma membrane protein, present on the apical membrane of human colonic and Caco-2 cells. It might have role in apical delivery. It is involved in the control of apical membrane trafficking in epithelial cells. It plays a crucial role in the growth of neurites during neuronal development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78881

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Delgrossi et al.
Journal of cell science, 110 ( Pt 18), 2207-2214 (1997-10-24)
To understand the molecular mechanisms underlying sorting of apical and basolateral membrane components in human intestinal epithelial cells, we have cloned the human homolog of rat syntaxin 3 and looked for its subcellular localization. Endogenous human syntaxin 3 was found
B Martín-Martín et al.
Journal of leukocyte biology, 65(3), 397-406 (1999-03-18)
Syntaxins are major components of vesicle trafficking and their pattern of expression depends on the cell type. Using reverse transcriptase-polymerase chain reaction (RT-PCR), cloning, and sequencing techniques, we have found that human neutrophils and neutrophil-differentiated HL-60 cells co-express syntaxins 1A
K Riento et al.
The Journal of biological chemistry, 275(18), 13476-13483 (2000-05-02)
The Sec1-related proteins bind to syntaxin family t-SNAREs with high affinity, thus controlling the interaction of syntaxins with their cognate SNARE partners. Munc18-2 is a Sec1 homologue enriched in epithelial cells and forms a complex with syntaxin 3, a t-SNARE
Frédéric Darios et al.
Nature, 440(7085), 813-817 (2006-04-07)
Growth of neurite processes from the cell body is the critical step in neuronal development and involves a large increase in cell membrane surface area. Arachidonic-acid-releasing phospholipases are highly enriched in nerve growth cones and have previously been implicated in
Poppy Datta et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(32), E4400-E4409 (2015-07-29)
Compartmentalization and polarized protein trafficking are essential for many cellular functions. The photoreceptor outer segment (OS) is a sensory compartment specialized for phototransduction, and it shares many features with primary cilia. As expected, mutations disrupting protein trafficking to cilia often

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service