Skip to Content
Merck
All Photos(5)

Key Documents

HPA001308

Sigma-Aldrich

Anti-USP14 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Deubiquitinating enzyme 14 antibody produced in rabbit, Anti-Ubiquitin carboxyl-terminal hydrolase 14 antibody produced in rabbit, Anti-Ubiquitin thioesterase 14 antibody produced in rabbit, Anti-Ubiquitin-specific-processing protease 14 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€540.00

€540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€540.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€540.00


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

EKESVNAKVLKDVKFPLMLDMYELCTPELQEKMVSFRSKFKDLEDKKVNQQPNTSDKKSSPQKEVKYEPFSFADDIGSNNCGYYDLQAVLTHQGRSSSSGHYVSWVKRKQDEWIKFDDDKVSIVTPEDILRLSGGGDWHIAYVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP14(9097)

Related Categories

General description

USP14 is made up of 494 amino acids containing a 45kDa catalytic domain at its N-terminus. It contains a ubiquitin-like (Ubl) domain at the N-terminus that associates with 19S regulatory particle (RP) 26S proteasome. This binding activates the deubiquitinating activity of the protein.

Immunogen

Ubiquitin carboxyl-terminal hydrolase 14 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

USP14 (ubiquitin specific peptidase 14) gene encodes a protein that belongs to the ubiquitin-specific processing (UBP) family of proteases. It is a proteasome-associated deubiquitinating enzyme (DUB) containing His and Cys catalytic domains. It releases ubiquitin from the proteasome targeted ubiquitinated proteins resulting in regeneration of ubiquitin at the proteasome. It is involved in the degradation of chemokine receptor CXCR4. CXCR4 is required for CXCL12-induced cell chemotaxis and its overexpression leads to cancer and inflammatory and neurodegenerative disorders. Under ER stress, It binds to α, a endoplasmic reticulum (ER) stress receptor, that is involved in unfolded protein response (UPR), a mechanism to control the accumulation of unfolded proteins within the (ER) lumen. Its overexpression leads to ER-associated degradation (ERAD) pathway inhibition.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77920

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marjelo A Mines et al.
The Journal of biological chemistry, 284(9), 5742-5752 (2008-12-25)
The chemokine receptor CXCR4 plays important roles in the immune and nervous systems. Abnormal expression of CXCR4 contributes to cancer and inflammatory and neurodegenerative disorders. Although ligand-dependent CXCR4 ubiquitination is known to accelerate CXCR4 degradation, little is known about counter
Min Hu et al.
The EMBO journal, 24(21), 3747-3756 (2005-10-08)
The ubiquitin-specific processing protease (UBP) family of deubiquitinating enzymes plays an essential role in numerous cellular processes. Mammalian USP14 (Ubp6 in yeast) is unique among known UBP enzymes in that it is activated catalytically upon specific association with the 26S
Atsushi Nagai et al.
Biochemical and biophysical research communications, 379(4), 995-1000 (2009-01-13)
Accumulation of unfolded proteins within the endoplasmic reticulum (ER) lumen induces ER stress. Eukaryotic cells possess the ER quality control systems, the unfolded protein response (UPR), to adapt to ER stress. IRE1alpha is one of the ER stress receptors and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service