Skip to Content
Merck
All Photos(1)

Key Documents

H5005

Sigma-Aldrich

4-Hydroxyphenylpyruvic acid

Grade II, 98-100%, crystalline

Synonym(s):

3-(4-Hydroxyphenyl)-2-oxopropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2COCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
2691632
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

type

Grade II

Assay

98-100%

form

crystalline

color

off-white to tan

mp

219-220 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

OC(=O)C(=O)Cc1ccc(O)cc1

InChI

1S/C9H8O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,10H,5H2,(H,12,13)

InChI key

KKADPXVIOXHVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Borowski et al.
Biochemistry, 43(38), 12331-12342 (2004-09-24)
Density functional calculations using the B3LYP functional has been used to study the reaction mechanism of 4-hydroxyphenylpyruvate dioxygenase. The first part of the catalytic reaction, dioxygen activation, is found to have the same mechanism as in alpha-ketoglutarate-dependent enzymes; the ternary
Nick J P Wierckx et al.
Journal of bacteriology, 190(8), 2822-2830 (2007-11-13)
The unknown genetic basis for improved phenol production by a recombinant Pseudomonas putida S12 derivative bearing the tpl (tyrosine-phenol lyase) gene was investigated via comparative transcriptomics, nucleotide sequence analysis, and targeted gene disruption. We show upregulation of tyrosine biosynthetic genes
Ute Metzger et al.
Journal of molecular biology, 404(4), 611-626 (2010-10-16)
CloQ is an aromatic prenyltransferase from the clorobiocin biosynthetic pathway of Streptomyces roseochromogenes var. oscitans. It is involved in the synthesis of the prenylated hydroxybenzoate moiety of the antibiotic, specifically catalyzing the attachment of a dimethylallyl moiety to 4-hydroxyphenylpyruvate. Herein
Alejandro Aranda et al.
International journal for parasitology, 36(3), 295-307 (2005-12-03)
Trypanosoma brucei procyclic forms possess three different malate dehydrogenase isozymes that could be separated by hydrophobic interaction chromatography and were recognized as the mitochondrial, glycosomal and cytosolic malate dehydrogenase isozymes. The latter is the only malate dehydrogenase expressed in the
Dingding Shao et al.
Acta tropica, 106(1), 9-15 (2008-02-12)
Macrophage migration inhibitory factor homologues have been identified from several genera of parasites, including Plasmodium, and have shown some functional similarities to the host molecule. It was hypothesized that MIF molecules can act as a regulator in host-parasite interaction in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service