Skip to Content
Merck
All Photos(4)

Key Documents

H4884

Sigma-Aldrich

Hippuryl-His-Leu acetate salt

Synonym(s):

N-Benzoyl-Gly-His-Leu

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€63.90
100 MG
€120.00
500 MG
€428.00
1 G
€738.00

€63.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€63.90
100 MG
€120.00
500 MG
€428.00
1 G
€738.00

About This Item

Empirical Formula (Hill Notation):
C21H27N5O5
CAS Number:
Molecular Weight:
429.47
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

€63.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

solubility

acetic acid: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)CNC(=O)c2ccccc2)C(O)=O

InChI

1S/C21H27N5O5.C2H4O2/c1-13(2)8-17(21(30)31)26-20(29)16(9-15-10-22-12-24-15)25-18(27)11-23-19(28)14-6-4-3-5-7-14;1-2(3)4/h3-7,10,12-13,16-17H,8-9,11H2,1-2H3,(H,22,24)(H,23,28)(H,25,27)(H,26,29)(H,30,31);1H3,(H,3,4)/t16-,17-;/m0./s1

InChI key

NSEVQQKNHZCIKL-QJHJCNPRSA-N

Gene Information

Related Categories

Amino Acid Sequence

NBz-Gly-His-Leu

Biochem/physiol Actions

Substrate for angiotensin converting enzyme.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mammalian Genome null
Matthew L Hemming et al.
The Journal of biological chemistry, 280(45), 37644-37650 (2005-09-13)
Human genetic data have associated angiotensin-converting enzyme (ACE) with Alzheimer disease (AD), and purified ACE has been reported to cleave synthetic amyloid beta-protein (Abeta) in vitro. Whether deficiency in ACE activity, arising from genetic alteration or pharmacological inhibition, can decrease
Neha Chaudhary et al.
Journal of food science and technology, 57(4), 1371-1381 (2020-03-18)
Controlled release of Emblicanin rich water soluble extract of Emblica officinalis (EEO) from the inner phase of water-in-oil-in-water type double emulsion (DE), during in vitro digestion and phagocytosis was investigated. It was observed that release of EEO (measured as total polyphenols
Jelena Kamilic et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 20(3), 170-179 (2009-03-04)
In humans, the insertion/deletion polymorphism in the angiotensin converting enzyme (ACE) gene accounts for half of the variance in plasma ACE activity. The deletion allele is associated with high plasma ACE activity, cardiovascular disease, and renal disease. In rat, a
Lucas Actis-Goretta et al.
FEBS letters, 555(3), 597-600 (2003-12-17)
It was determined that flavan-3-ols and procyanidins have an inhibitory effect on angiotensin I converting enzyme (ACE) activity, and the effect was dependent on the number of epicatechin units forming the procyanidin. The inhibition by flavan-3-ols and procyanidins was competitive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service