Skip to Content
Merck
All Photos(1)

Key Documents

H4413

Sigma-Aldrich

Hydroxyfasudil hydrochloride hydrate

≥98% (HPLC), solid

Synonym(s):

1-(1-Hydroxy-5-isoquinolinesulfonyl)homopiperazine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€271.00
25 MG
€1,020.00

€271.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€271.00
25 MG
€1,020.00

About This Item

Empirical Formula (Hill Notation):
C14H17N3O3S · HCl · xH2O
CAS Number:
Molecular Weight:
343.83 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€271.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

composition

The two peaks show the equilibrim between the two tautomers

color

white

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].Oc1nccc2c(cccc12)S(=O)(=O)N3CCCNCC3

InChI

1S/C14H17N3O3S.ClH/c18-14-12-3-1-4-13(11(12)5-7-16-14)21(19,20)17-9-2-6-15-8-10-17;/h1,3-5,7,15H,2,6,8-10H2,(H,16,18);1H

InChI key

XWWFOUVDVJGNNG-UHFFFAOYSA-N

Application

Hydroxyfasudil hydrochloride hydrate has been used as a ROCK inhibitor to study the role of different signaling pathways in the chemokines CCL19 and CCL21, the CCR7 ligands mediated effects on migratory and prosurvival, in CLL cells.[1]

Biochem/physiol Actions

Hydroxyfasudil possessantispastic action. It is an active metabolite of fasudil. Hydroxyfasudil might enhance the dynamics of blood flow and prevent damage caused by neutrophils.[2]
Rho-kinase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological profile of hydroxy fasudil as a selective rho kinase inhibitor on ischemic brain damage
Satoh SI, et al.
Life Sciences, 69(12), 1441-1453 (2001)
Analysis of migratory and prosurvival pathways induced by the homeostatic chemokines CCL19 and CCL21 in B-cell chronic lymphocytic leukemia
Cuesta-Mateos C, et al.
Experimental Hematology, 38(9), 756-764 (2010)
Ghulam Mohammad et al.
Annals of clinical and laboratory science, 48(2), 137-145 (2018-04-22)
To investigate the effects of blocking Rho kinase pathway on the expression of inflammatory signaling pathways in the retina of diabetic mice and in human retinal Müller glial cells stimulated with high-glucose to replicate hyperglycemia. Retinas from diabetic mice and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service