Skip to Content
Merck
All Photos(1)

Key Documents

H4128

Sigma-Aldrich

11β-Hydroxytestosterone

crystalline

Synonym(s):

11β,17β-Dihydroxyandrost-4-en-3-one, 4-Androstene-11β,17β-diol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystalline

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

light yellow

SMILES string

CC12CC(O)C3C(CCC4=CC(=O)CCC34C)C1CCC2O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Structure-activity relationships of 11-substituted testosterone derivatives as progesterone agonists, and their effect on growth and differentiation of human decidual cells.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Leitz et al.
General and comparative endocrinology, 58(3), 471-477 (1985-06-01)
Minced testes of the Siamese fighting fish Betta splendens were incubated with [14C]androstenedione at 27 degrees C for 15, 30, 60, and 120 min. The metabolic products were characterized by paper and thin-layer chromatography, derivative formation, and eventually by crystallization
S T Lee et al.
General and comparative endocrinology, 99(1), 41-49 (1995-07-01)
The steroidogenic potential of gonadal tissues of the protogynous grouper (Epinephelus tauvina) was examined before and after sex inversion with 17 alpha-methyltestosterone. Incubations of gonadal tissues of the fish with [3H]testosterone resulted in the biosynthesis of 5 beta-androstane-3 beta, 17
P M Rosenblum et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 82(4), 659-665 (1985-01-01)
Highly specific antisera for 11-keto- and 11 beta-hydroxytestosterone have been raised in sheep. Assay systems for the simultaneous measurement of 11-ketotestosterone, 11 beta-hydroxytestosterone, testosterone, progesterone and estradiol-17 beta were validated for Ictalurus nebulosus plasma and Carassius auratus serum. In males
F Ollevier et al.
General and comparative endocrinology, 61(2), 214-228 (1986-02-01)
The O-pentafluorobenzyloxime (OPFB)-heptafluorobutyrylester (HFB) derivatives of hemolymph steroid extracts from both male and female Astacus leptodactylus were subjected to negative ion chemical ionization and capillary gas chromatography-mass spectrometry (NCI/GC-MS). Five nonecdysteroid steroids, namely pregnenolone, 17 alpha-hydroxypregnenolone, testosterone, cholesterol, and 6
R Schulz
General and comparative endocrinology, 64(2), 312-319 (1986-11-01)
Male rainbow trout show high plasma androgen levels beginning with the period of full spermatogenesis until the end of spermiation. The difficulties in explaining the steroid levels in regard to the concomitant changes of the plasma GTH concentrations prompted investigations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service