Skip to Content
Merck
All Photos(1)

Key Documents

H3661

Sigma-Aldrich

4′-Hydroxydiclofenac

≥98% (HPLC)

Synonym(s):

4′-Hydroxy Diclofenac, [2-(2,6-Dichloro-4-hydroxy-phenylamino)phenyl]acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11Cl2NO3
CAS Number:
Molecular Weight:
312.15
Beilstein:
4198042
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.47
Product H3661 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥98% (HPLC)

packaging

vial of 5 mg

storage temp.

−20°C

SMILES string

OC(=O)Cc1ccccc1Nc2c(Cl)cc(O)cc2Cl

InChI

1S/C14H11Cl2NO3/c15-10-6-9(18)7-11(16)14(10)17-12-4-2-1-3-8(12)5-13(19)20/h1-4,6-7,17-18H,5H2,(H,19,20)

InChI key

KGVXVPRLBMWZLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4′-Hydroxydiclofenac is an important metabolite of aceclofenac, seen mainly in rats.[1] It is the important oxidative metabolite found in urine.[2]
CYP2C9 metabolite of diclofenac.

Application

4′-Hydroxydiclofenac has been used to investigate the effect of gentiopicroside (GE) on human cytochrome P450 enzymes in vitro.[3] It has also been used as an enzyme marker to study its metabolic activities in different in vitro liver models.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic profiling of human long-term liver models and hepatic clearance predictions from in vitro data using nonlinear mixed-effects modeling
Kratochwil NA, et al.
The AAPS Journal, 19(2), 534-550 (2017)
Metabolism of Drugs and Other Xenobiotics (2012)
In vitro inhibition and induction of human liver cytochrome P450 enzymes by gentiopicroside: potent effect on CYP2A6
Deng Y, et al.
Drug Metabolism and Pharmacokinetics (2013)
Lu Liu et al.
Pharmaceutical biology, 55(1), 1868-1874 (2017-06-16)
Dihydromyricetin (DHM) is the most abundant and active flavonoid component isolated from Ampelopsis grossedentata (Hand-Mazz) W.T. Wang (Vitaceae) and it possesses numerous pharmacological activities. However, whether DHM affects the activity of human liver cytochrome P450 (CYP) enzymes remains unclear. The
Baodong Ding et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(8), 913-918 (2020-02-07)
Pachymic acid is a wildly used traditional Chinese medicine with various pharmacological features. It also exists in many drugs which are wildly used in pediatric.The effect of pachymic acid on the activity of eight major CYP isoforms was investigated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service