Skip to Content
Merck
All Photos(1)

Key Documents

H1667

Sigma-Aldrich

Hypothemycin

from Hypomyces trichothecoides, ≥98% (HPLC)

Synonym(s):

3H-Oxireno[k][2]benzoxacyclotetradecin-5,11(2H,4H)-dione,1a,8,9,15b-tetrahydro-3,4,12-trihydroxy-14-methoxy-9-methyl-,(1aR,3S,4S,6Z,9S,15bR)-, NSC354462

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€206.00
1 MG
€473.00

€206.00


Please contact Customer Service for Availability


Select a Size

Change View
250 μG
€206.00
1 MG
€473.00

About This Item

Empirical Formula (Hill Notation):
C19H22O8
CAS Number:
Molecular Weight:
378.37
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€206.00


Please contact Customer Service for Availability

biological source

Hypomyces trichothecoides

Quality Level

Assay

≥98% (HPLC)

solubility

DMSO: soluble
acetone: soluble
deionized water: insoluble

storage temp.

−20°C

SMILES string

COc1cc(O)c2C(=O)O[C@@H](C)CC=CC(=O)[C@@H](O)[C@@H](O)CC3OC3c2c1

InChI

1S/C19H22O8/c1-9-4-3-5-12(20)17(23)14(22)8-15-18(27-15)11-6-10(25-2)7-13(21)16(11)19(24)26-9/h3,5-7,9,14-15,17-18,21-23H,4,8H2,1-2H3/b5-3-/t9-,14-,15+,17+,18+/m0/s1

InChI key

SSNQAUBBJYCSMY-KNTMUCJRSA-N

Biochem/physiol Actions

Hypothemycin, one of the highly oxygenated analogues in the group of 14-membered resorcylic acid lactones (RAL), has minor antifungal and cytotoxic activity and exhibits an in vitro anti-malarial activity with an IC50 of 2.2 μg/mL.
Hypothemycin, one of the highly oxygenated analogues in the group of 14-membered resorcylic acid lactones (RAL), has minor antifungal and cytotoxic activity and exhibits an in vitro anti-malarial activity with an IC50 of 2.2 μg/mL. Hypothemycin is also reported to selectively and irreversibly inhibit protein kinases that contain a conserved cysteine residue (Cys166) that is located within the ATP-binding domain. Though this group accounts for less then 10% of all identified kinases, there are several targets implicated in aberrant cellular proliferation such as ERKs, MEK, FMS-like tyrosine kinase protein (FLT), and platelet-derived growth factor receptors (PDGFR). In cell culture, hypothemycin displays potent cytotoxicity against cancer cell lines that are dependent on certain activating kinase mutations. Additionally, hypothemycin demonstrates significant tumor growth inhibition in at least three separate murine xenograft models. Hypothemycin also inhibits the production of several cytokines such as IL2, IL6, IFNγ, and TNFα.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marilù Fanelli et al.
Frontiers in oncology, 10, 331-331 (2020-03-27)
Treatment of high-grade osteosarcoma, the most common malignant tumor of bone, is largely based on administration of cisplatin and other DNA damaging drugs. Altered DNA repair mechanisms may thus significantly impact on either response or resistance to chemotherapy. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service