H145
(±)-7-hydroxy-dpat hydrobromide
Synonym(s):
(±)-2-Dipropylamino-7-hydroxy-1,2,3,4-tetrahydronaphthalene hydrobromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H25NO · HBr
CAS Number:
Molecular Weight:
328.29
MDL number:
UNSPSC Code:
12352200
SMILES string
Br.CCCN(CCC)C1CCc2ccc(O)cc2C1
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Eriko Ikeda et al.
Molecular pharmacology, 83(5), 959-967 (2013-02-23)
The dopamine D3 receptor (DRD3) in the ventral striatum is thought to influence motivation and motor functions. Although the expression of DRD3 in the ventral striatum has been shown to exhibit 24-hour variations, the mechanisms underlying the variation remain obscure.
Y Iris Chen et al.
The European journal of neuroscience, 34(5), 800-815 (2011-09-08)
Chronic use of cocaine is associated with lasting alterations in brain metabolism, circuitry, and receptor properties. We used neuroimaging with pharmacological magnetic resonance imaging to assess alterations in response to cocaine (0.5 mg/kg) in animals trained to self-administer cocaine on
Mikhail N Koffarnus et al.
Psychopharmacology, 203(2), 317-327 (2008-09-23)
Previous research has found the stimulus effects of dopamine D2- and D3-preferring agonists difficult to distinguish in drug discrimination studies. Antagonism studies suggest that the stimulus effects of both types of agonists may be mediated primarily through D2 receptors. The
Gavin D Phillips et al.
Psychopharmacology, 203(1), 161-173 (2008-10-25)
Two issues were addressed regarding the effects of amygdala dopamine manipulations on associative learning: first, an apparent contradiction between the effects of post- vs. pre-session dopaminergic manipulations and second, the ability of dopaminergic infusions to affect association formation vs. its
Rosario Sánchez-Pernaute et al.
Neurobiology of disease, 27(2), 220-227 (2007-06-26)
A growing body of evidence indicates a role for D(3) receptors in l-DOPA-induced dyskinesias. This involvement could be amenable to non-invasive in vivo analysis using functional neuroimaging. With this goal, we examined the hemodynamic response to the dopamine D(3)-preferring agonist
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service