H0162
Hide Powder
from bovine hide, protease substrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine hide
form
powder
Preparation Note
Non-chromated
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the science of food and agriculture, 99(6), 2855-2864 (2018-11-20)
Fruits present high concentrations of bioactive compounds that are beneficial to health due to their antioxidant properties. New alternatives to vegetable oils with such properties have been studied. We determined the chemical compounds of Byrsonima cydoniifolia A. Juss. fruits in
Infection and immunity, 76(5), 1825-1836 (2008-02-06)
Secretion of effector molecules is one of the major mechanisms by which the intracellular human pathogen Legionella pneumophila interacts with host cells during infection. Specific secretion machineries which are responsible for the subfraction of secreted proteins (soluble supernatant proteins [SSPs])
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service