Skip to Content
Merck
All Photos(1)

Key Documents

GW22770

Sigma-Aldrich

Anti-MAPK7 (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€269.00

€269.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€269.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€269.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22770

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK7(5598)

Immunogen

Immunogen Sequence: GI # 20986497, sequence 341-570
Recombinant mitogen-activated protein kinase 7 isoform 1

Application

Anti-MAPK7 (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Mitogen-activated protein kinase 7 (also known as MAP kinase 7) is an enzyme encoded by the MAPK7 gene in humans and is mapped to chromosome 17 band p11.2. It belongs to mitogen-activated protein kinase (MAPK) family and regulates a wide variety of cellular processes such as proliferation, differentiation, necrosis, apoptosis and degeneration. MAPK7 is involved in degeneration of nucleus pulposus. Expression of MAPK7 may act as a valuable indicator for predicting lung cancer risk in smokers.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuman Qiu et al.
Mutagenesis, 28(5), 561-567 (2013-06-28)
Mitogen/extracellular signal-regulated kinase-5 (MEK5)/extracellular signal-regulated protein kinase-5 (ERK5) pathway plays a pro-oncogenic role in tumourigenesis by anticell apoptosis, promoting cell proliferation and differentiation in response to extracellular stimuli. As overexpressed MEK5/ERK5 is involved in the development of lung cancer, we
Weiguo Liang et al.
Biochemical and biophysical research communications, 449(4), 466-470 (2014-05-27)
Extracellular-signal-regulated kinase 5 (ERK5) is a member of the mitogen-activated protein kinase (MAPK) family and regulates a wide variety of cellular processes such as proliferation, differentiation, necrosis, apoptosis and degeneration. However, the expression of ERK5 and its role in degenerated
Assignment of big MAP kinase (PRKM7) to human chromosome 17 band p11.2 with somatic cell hybrids.
S M Purandare et al.
Cytogenetics and cell genetics, 83(3-4), 258-259 (1999-03-11)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service