Skip to Content
Merck
All Photos(1)

Key Documents

GERPN3006

ECL Gold Buffer

Cytiva RPN3006, pack of 1 ea

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,630.00

€1,630.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€1,630.00

About This Item

UNSPSC Code:
41105300
NACRES:
NA.31

€1,630.00


Please contact Customer Service for Availability

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Cytiva RPN3006

Related Categories

General description

ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Okumura et al.
The Journal of biological chemistry, 273(31), 19469-19475 (1998-07-25)
The phospholipase A2 (PLA2) inhibitor PLIbeta, purified from the blood plasma of Chinese mamushi snake (Agkistrodon blomhoffii siniticus), is a 160-kDa trimer with three 50-kDa subunits; and it inhibits specifically the enzymatic activity of the basic PLA2 from its own
Cory M Robinson et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(25), 9667-9672 (2006-06-13)
Enterohemorrhagic Escherichia coli (EHEC) 0157:H7 is a food-borne pathogen that can cause bloody diarrhea and, occasionally, acute renal failure as a consequence of Shiga toxin (Stx) production by the organism. Stxs are potent cytotoxins that are lethal to animals at
H Nonogaki et al.
Plant physiology, 123(4), 1235-1246 (2000-08-12)
Endo-beta-mannanase (EC 3.2.1.78) is involved in hydrolysis of the mannan-rich cell walls of the tomato (Lycopersicon esculentum Mill.) endosperm during germination and post-germinative seedling growth. Different electrophoretic isoforms of endo-beta-mannanase are expressed sequentially in different parts of the endosperm, initially
T Yugawa et al.
Journal of virology, 72(3), 2272-2279 (1998-03-14)
We previously reported (N. Watanabe, M. Nishi, Y. Ikawa, and H. Amanuma, J. Virol. 65:132-137, 1991) that the mutant Friend spleen focus-forming virus (F-SFFV(MS)), which encodes a mutant gp55 membrane glycoprotein with an ecotropic env gp70 sequence, was nonpathogenic. Here
D S Li et al.
The Journal of general virology, 75 ( Pt 7), 1713-1722 (1994-07-01)
Libraries of cDNA were generated from polyadenylated RNAs derived from Marek's disease virus (MDV)-transformed cell lines by directional cloning of oligo-(dT)-primed cDNAs in lambda gt22A. Analysis of the libraries for viral sequences showed that a number of cDNA clones originated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service