Skip to Content
Merck
All Photos(1)

Key Documents

GERPN2124

Amersham ECL Western Blotting Reagent Pack

Cytiva RPN2124, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

Anti-rabbit IgG, peroxidase linked whole antibody (donkey): Colorless Anti-mouse IgG, peroxidase linked whole antibody (sheep): Colorless ECL membrane blocking agent: White

feature

wetted part: no

packaging

pack of 1 kit

manufacturer/tradename

Cytiva RPN2124

storage temp.

2-8°C

General description

Each pack contains the following reagents:

Anti-mouse IgG horseradish peroxidase conjugate, 100 μL
Anti-rabbit IgG horseradish peroxidase conjugate, 100 μL
ECL membrane blocking agent, 5 g.

Sufficient for at least 10 miniblots.

Application

Amersham ECL western blotting reagent pack has been used in high sensitivity western blotting.[1]

Features and Benefits

  • Contains reagents for membrane blocking and secondary detection of membrane-bound mouse and rabbit primary antibodies.
  • Designed for use with nitrocellulose or PVDF membranes.
  • Detection reagents are not included, but the system is optimized for use with Amersham ECL Western Blotting Reagents, and Amersham ECL Prime.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Amersham is a trademark of Cytiva
ECL is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Factor Inhibiting HIF Asparaginyl Hydroxylase Regulates Oxidative Metabolism and Accelerates Metabolic Adaptation to Hypoxia.
Sim J, et al.
Cell Metabolism, 27(4), 898-913 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service