Skip to Content
Merck
All Photos(1)

Documents

GEE79000Y

DNA Polymerase, Thermo Sequenase

1,000 Units, Cytiva E79000Y

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106314

feature

1,000 Units

manufacturer/tradename

Cytiva E79000Y

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates.

  • Uniform and easy-to-read band patterns.
  • Thermostable enzyme.


Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates. This results in extremely uniform and easy-to-read sequence band patterns. This property, in addition to thermostability, makes Thermo Sequenase an excellent choice for cycle sequencing where template quantity limitations preclude the use of T7 Sequenase DNA polymerase. The properties of Thermo Sequenase also enable its use in many SNP genotyping applications where primer extension is employed.

Features and Benefits

  • Uniform and easy-to-read band patterns.
  • Better incorporation of ddNTPs helps avoid secondary structures that can cause unreadable sequences
  • Read-through of difficult sequences (e.g., GC-rich), due to the ability of thermostable enzymes to function at elevated temperature used for sequencing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service