Skip to Content
Merck
All Photos(1)

Key Documents

GEE70073X

Exonuclease I, 10 U/μL

Cytiva E70073X, pack of 5000 units

Synonym(s):

Exonuclease I Enzyme

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€757.00

€757.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€757.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.53

€757.00


Please contact Customer Service for Availability

biological source

Escherichia coli

packaging

pack of 5000 units

manufacturer/tradename

Cytiva E70073X

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Acts specifically on single-stranded DNA degrading it processively in the 3′- to 5′-direction, producing 5′-mononucleotides.
Exonuclease I (Exo I) from E. coli is an Mg2+ ion dependent exonuclease.[1]

Application

Exonuclease I has been used to quantify the expression of 56 genes in each F2 XY gonad.[2]

Biochem/physiol Actions

Exonuclease I (Exo I) from E. coli can degrade single-stranded DNA (ssDNA) from 3′ to 5′. It participates in various DNA repair systems, like, RecBCD-dependent homologous recombination, methyl-dependent mismatch repair and genomic tandem repeat maintenance.[1]

Features and Benefits

  • Elimination of residual single-stranded DNA containing a 3′-terminus.
  • Measuring endonucleolytic cleavage of covalently closed circular (ccc) ssDNA.
  • Measuring DNA helicase activity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and Biochemical Studies of a Moderately Thermophilic Exonuclease I from Methylocaldum szegediense
Fei L, et al.
Testing, 10(2), e0117470-e0117470 (2015)
Elucidation of the transcription network governing mammalian sex determination by exploiting strain-specific susceptibility to sex reversal
Munger S C, et al.
Genes & Development, 23(21), 2521-2536 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service