Skip to Content
Merck
All Photos(1)

Key Documents

GE90-1004-08

Sephacryl S-300 HR, XK 16/100

Sign Into View Organizational & Contract Pricing

Select a Size

GE90-1004-08
€4,820.00

€4,820.00


Please contact Customer Service for Availability


Select a Size

Change View
GE90-1004-08
€4,820.00

About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

€4,820.00


Please contact Customer Service for Availability

manufacturer/tradename

Cytiva

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Separation options for Size Exclusion Chromatography with Sephacryl from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service