Skip to Content
Merck
All Photos(1)

Key Documents

GE80-1106-58

Grommet for MultiPhor II electrophoresis unit

pack of 2 ea

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€62.60

€62.60


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€62.60

About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

€62.60


Please contact Customer Service for Availability

grade

electrophoresis grade

packaging

pack of 2 ea

manufacturer/tradename

Cytiva 80110658

Related Categories

General description

Grommets are connected to the inlet and outlet tubes of the cooling plate which can then be connected to a thermostatic ciculator such as MultiTemp III.

Legal Information

MultiPhor is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service