Skip to Content
Merck
All Photos(1)

Key Documents

GE29275879

Capto HiRes

S 10/100

Synonym(s):

Capto HiRes Resin, Chromatography Resin, High Resolution Capto

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€6,010.00

€6,010.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€6,010.00

About This Item

UNSPSC Code:
23151817
NACRES:
NB.21

€6,010.00


Please contact Customer Service for Availability

manufacturer/tradename

Cytiva 29275879

Looking for similar products? Visit Product Comparison Guide

General description

Capto HiRes Q and Capto HiRes S are prepacked columns for high-resolution purification of proteins and other biomolecules by ion exchange chromatography. They are well-suited for the preparation of highly pure proteins for structural studies and for protein analysis.


Capto HiRes ion exchange chromatography resin


The chromatography resins packed in Capto HiRes Q and Capto HiRes S ion exchange columns are based on a high-flow agarose base matrix and a small bead size (average bead size 9 μm).

Capto HiRes Q and Capto HiRes S are strong anion and strong cation exchangers, respectively with ion exchange properties that remain constant during chromatography within the recommended pH range of 2 to 12.

A high-performance IEX column format


Capto HiRes Q and Capto HiRes S IEX resins are prepacked in high-performance Tricorn glass columns of two different sizes. The columns have fittings for simple connection to ÄKTA chromatography systems or equivalent high-performance systems. All column parts are biocompatible.

High-resolution protein purification


In many research areas, for example in structural biology using X-ray crystallography or cryo-electron microscopy (cryo-EM), obtaining homogeneous size and charge of biomolecules is crucial for the elucidation of their structures. High-resolution separation of samples based on their charge properties is essential to secure sample charge homogeneity and success of the study. Capto HiRes columns separate proteins and biomolecules that are very close in net charge, which makes these columns great for these applications.

Protein analysis


Capto HiRes ion exchange columns can be used for protein analysis, in applications such as characterization of target protein charge variants.

Capto Q HiRes and Capto S HiRes replace MonoBeads columns


A separation that worked on a Mono Q or Mono S column may be performed on a Capto HiRes Q or Capto HiRes S column with little modification or optimization. Similar resin selectivity and slightly improved resolution can be expected with the Capto HiRes columns while using the same experimental conditions. The similar selectivity of the two columns ensures a smooth transition even for quality control (QC) applications.

Features and Benefits

  • High resolution columns—for lab scale ion exchange chromatography.
  • Suitable for preparing highly pure proteins—essential in structural studies to perform techniques such as cryo-electron microscopy or protein crystallography.
  • Suitable for protein analysis in research or quality control—for example, for change variants analysis, or for studying homogeneity in expression.
  • High quality column—excellent reproducibility, durability, and high chemical and pH stabilities.
  • Replacement for Mono Qand Mono Scolumns—delivering similar or improved resolution, with easy transition from one column platform to the other.

Legal Information

Capto is a trademark of Cytiva

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service