GE29-0362-26
Superose® 6 3.2/300
Cytiva 29-0362-26, 13 μm particle size
About This Item
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 29-0362-26
availability
available only in USA, Canada and Puerto Rico
matrix
agarose
matrix active group
agarose phase
particle size
13 μm
separation technique
size exclusion (SEC)
General description
Features and Benefits
- Achieve high-resolution separations across exceptionally broad molecular-weight ranges with Superose gel filtration medium.
- Precision columns (PC) for micropurification and analysis: sample volumes of 4 - 50 μL.
- Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds.
- New hardware design for convenient direct connection to high performance chromatography systems such as ÄKTA design.
Storage and Stability
Analysis Note
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service