Key Documents
GE28-9889-47
Sigma-Aldrich
XK 16/100 Column
Select a Size
1 EA
€952.00
€952.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€952.00
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
€952.00
Please contact Customer Service for Availability
material
polypropylene column
packaging
1 ea of
General description
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
Features and Benefits
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Column Packing and Preparation for Size Exclusion Chromatography