Skip to Content
Merck
All Photos(1)

Key Documents

GE28908366

HiTrap® IgG Sepharose FF

pkg of 5 × 5 mL, Cytiva, 28-9083-66, 90 μm avg. part. size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NB.21
Product GE28908366 is not currently sold in your country. Contact Technical Service

packaging

pkg of 5 × 5 mL

manufacturer/tradename

28-9083-66
Cytiva

avg. part. size

90 μm

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service