Skip to Content
Merck
All Photos(1)

Key Documents

GE28-4064-14

Sigma-Aldrich

Tricorn 10/50 Column

Synonym(s):

Tricorn column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€758.00

€758.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€758.00

About This Item

UNSPSC Code:
41121800
NACRES:
SB.52

€758.00


Please contact Customer Service for Availability

Request a Bulk Order

technique(s)

LPLC: suitable

General description

Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Tricorn columns include a glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock, and filter holder.

Features and Benefits

Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes efficient column packing and preparation for affinity chromatography of antibodies.

This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

Protocols

This page shows how to purify histidine-tagged proteins using TALON Superflow from Cytiva.

This page shows how to separate IgG antibodies by affinity chromatography using Protein G Sepharose 4 Fast Flow from Cytiva.

This page describes efficient column packing and preparation for affinity chromatography of GST-tagged proteins using Cytiva products.

This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service