Skip to Content
Merck
All Photos(1)

Key Documents

GE27-1006-03

A 5′-TP Na Salt

Cytiva 27-1006-03, pack of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.52
Pricing and availability is not currently available.

form

solid

packaging

pack of 25 g

manufacturer/tradename

Cytiva 27-1006-03

color

White

shipped in

dry ice

storage temp.

−20°C

General description

High purity rNTPs are easy-to-use, time-saving solutions for in vitro transcription assays and/or reactions requiring Ribonucleoside Triphosphates (rNTPs) of high purity. This solution can be used for in vitro transcription or for reactions requiring rNTPs of high purity.

Features and Benefits

  • Each lot is tested for ribonuclease contamination to ensure success in your application.
  • Supplied in purified water as 100 mM solutions (pH 7.5).
  • Extensively purified to yield a minimum of 99% in triphosphate form for solution (98% for lyophilized) to enhance incorporation and yield optimal experimental results.
  • Available either as individual products or in a convenient set containing ATP, CTP, GTP, and UTP in separate tubes.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service