Key Documents
GE25-9005-98
Autoscreen A, 96-Well Plate Pack
Select a Size
10 X 96 WELLS
€1,040.00
€1,040.00
Please contact Customer Service for Availability
Select a Size
Change View
10 X 96 WELLS
€1,040.00
About This Item
UNSPSC Code:
41106305
NACRES:
NA.52
€1,040.00
Please contact Customer Service for Availability
manufacturer/tradename
Cytiva
packaging
pkg of (10 x 96-well plates)
General description
AutoScreen 96-Well Plate consists of a 96-well filter plate containing DNA Grade Sephadex G-50 for purification of sequencing reactions prior to analysis on ABI sequencers and can be used for other size exclusion applications.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)