Skip to Content
Merck
All Photos(1)

Documents

GE17548701

Cytodex® 1 Gamma Microcarriers

sterile, dry powder, Cytiva, pkg of 30 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161502
NACRES:
NC.01

sterility

sterile

feature

dry powder

packaging

pkg of 30 g

manufacturer/tradename

17548701
Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

Microcarrier for general use in sterilized dry powder form.

  • Gamma-sterilized and ready to use for quick culture start-up.
  • Delivered in a convenient, single-use container system for easy transfer to various cell culture vessels.
  • Supplied dry to save storage space and facilitate transportation.
  • Enable culturing of anchorage-dependent cells in single-use bioreactor to eliminate the need for cleaning and cleaning validation.
  • Supported by extensive product documentation to facilitate regulatory filing.
  • Serum-free function testing to ensure performance


A general-purpose microcarrier
Sterilized Cytodex 1 Gamma is a general-purpose microcarrier particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service