Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5481-51

HiTrap® Capto Core 700

Cytiva 17-5481-51, pack of 5 × 1 mL

Synonym(s):

Capto Core 700 Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56
Product GE17-5481-51 is not currently sold in your country. Contact Technical Service

ligand

octylamine

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-5481-51

parameter

5 bar

column I.D.

7 mm

matrix

highly cross-linked agarose

avg. part. size

75 (d₅₀v is the median particle size of the cumulative volume distribution)

cleaning

3-14(pH interval where the medium can be subjected to, for cleaning- or sanitization-in-place.)

working range

2-14(pH interval where the medium can be operated without significant change in function.)

capacity

~13 mg binding capacity (ovalbumin)

suitability

suitable for bioprocess medium

Looking for similar products? Visit Product Comparison Guide

General description

Dynamic binding capacities were measured at 10% breakthrough with a residence time of 3 min (1.6 ml/min = 200 cm/h) in HiScreen columns. Buffer: 20 mM Tris-HCl, 150 mM NaCl, pH 7.5.
HiTrap Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess resin for intermediate purification and polishing of viruses and other large biomolecules. Capto Core 700 is a novel, layered bead chromatography resin consisting of a porous outer layer and ligand-activated core. The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged. These internalized ligands bind impurities strongly over a wide range of pH and salt concentration. The pores of the bead outer layer prevent large targets (Mr > 700 000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured.

Features and Benefits

  • High capacity and productivity in flowthrough mode.
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification.
  • Wide operational window of pH and conductivity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service