Skip to Content
Merck
All Photos(2)

Key Documents

GE17-5474-15

HiScreen MabSelect SuRe LX

Cytiva 17-5474-15

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,340.00

€1,340.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€1,340.00

About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

€1,340.00


Please contact Customer Service for Availability

packaging

pkg of 4.7 mL

manufacturer/tradename

Cytiva 17-5474-15

storage condition

(20% Ehtanol)

parameter

44 psi

bed size

7.7 mm × 100 mm

bed volume

4.7 mL

storage temp.

2-8°C

Related Categories

General description

HiScreen MabSelect SuRe LX is prepacked with MabSelect SuRe LX and is part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies when a high binding capacity is needed.

Application

The small column volume of 4.7 mL and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up. Process fluid velocities can be applied, since the 10 cm bed height.

If necessary, two columns can easily be connected in series to give a bed height of 20 cm.

Media prepacked in HiScreen MabSelect SuRe LX columns are also available in other formats and as bulk packs, which makes it possible to use the same media for development work, pilot studies, and routine production.

Features and Benefits

  • The alkali-tolerant recombinant Protein A ligand of MabSelect Sure LX allows cleaning-in-place (CIP) and the medium gives increased binding capacity.
  • Prepacked, ready-to-use columns for convenient process development.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Free MabSelect SuRe Ligand
"MabSelect SuRe Ligand Restricted License" and "Cys-rProtein A Ligand Restricted License" are protected by the following patents and equivalent patents and patent applications in other countries: US 6,399,750 and EP 1123389. A free, non-transferable limited license to use this product for internal analytical purposes only accompanies the purchase of the product from a Cytiva company and its licensed distributors. Any other use will require a separate license from a Cytiva company.
HiScreen is a trademark of Cytiva
MabSelect is a trademark of Cytiva
MabSelect SuRe is a trademark of Cytiva
SuRe is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service