Skip to Content
Merck
All Photos(1)

Documents

GE17-5186-01

Sigma-Aldrich

SOURCE 15PHE 4.6/100 PE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

ligand

phenyl

material

polypropylene column

packaging

pkg of 1 ea

storage condition

(20% Ethanol)

parameter

580 psi

bed size

4.6 mm × 100 mm

bed volume

1.7 mL

column I.D.

4.6 mm

matrix

polymeric
polystyrene/divinylbenzene

avg. part. size

15 μm

cleaning

1-14

working range

2-12

suitability

suitable for bioprocess medium

separation technique

hydrophobic interaction (HIC)

General description

SOURCE 15PHE 4.6/100 PE is a Tricorn high performance column pre-packed with SOURCE 15PHE.

Application

SOURCE 15PHE 4.6/100 PE high capacity makes it an excellent choice for preparative hydrophobic interaction purifications at laboratory scale. In addition, its longer bed height provides increased resolution. The column is also useful for optimization studies during scale-up.

Features and Benefits

  • High resolution separations at high loads and high fLow rates.
  • Wide range of selectivities.
  • Reproducible quality.
  • Excellent scalability.
  • High chemical and physical stability.
  • Low backpressure.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SOURCE is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page discusses how to do a purification with SOURCE™ media from Cytiva.

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service