GE17372212
HiTrap® Capto™ AVB
pkg of 1 × 5 mL, Cytiva 17372212
Synonym(s):
Affinity Purification Column
Select a Size
€1,870.00
Select a Size
About This Item
€1,870.00
Recommended Products
packaging
pkg of 1 × 5 mL
manufacturer/tradename
Cytiva 17372212
parameter
20 mL/min max. flow rate
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Efficient purification of AAV of several subclasses by affinity chromatography
- Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
- High selectivity and excellent scalability
Legal Information
related product
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service