Skip to Content
Merck
All Photos(1)

Documents

GE17-1273-01

SOURCE 30S

Cytiva 17-1273-01, pack of 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

sulfonate group

description

Ion Exchanger Type (value)

packaging

pack of 50 mL

manufacturer/tradename

Cytiva 17-1273-01

matrix

polystyrene/divinylbenzene

particle size

30 μm

cleaning in place

1-14

working range

2-13

suitability

suitable for bioprocess medium

Related Categories

General description

SOURCE 30S is a polymeric, strong cation exchanger designed for intermediate purification and large-scale polishing.

Application

SOURCE 30S is a synthetic high performance, preparative, chromatography medium, based on a 30 μm monosized, rigid polystyrene/divinyl benzene polymer matrix It is modified with sulphonate (S) strong cation exchange groups. SOURCE media have excellent physical and chemical characteristics, allowing high flow rates and consistent performance at both laboratory and process scales.

SOURCE 30S is suitable for intermediate purification steps in industrial processes when large volumes of partially purified material need to be processed. A high degree of purification can be obtained with high productivity.

Through the combination of high quality assurance standards and a patented manufacturing process, particle structure is consistent both bead-to-bead and batch-to batch. As member of the BioProcess media range, SOURCE 30S meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Mono-sized 30 μm rigid beads gives Low back pressure and high flow rates allowing for high productivity
  • Strong cation exchanger designed for high resolution polishing purification of Proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

SOURCE is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows Hydrophobic Interaction Chromatography (HIC) in a purification strategy.

Protocols

In this section the practical aspect of Reverse Phased Chromatography ( RPC) is discussed including media and column selection and eluent selection and preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service