Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1186-01

RESOURCE PHE

Cytiva 17-1186-01, pack of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,240.00

€1,240.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€1,240.00

About This Item

UNSPSC Code:
13111000
NACRES:
NA.56

€1,240.00


Please contact Customer Service for Availability

ligand

phenyl

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 17-1186-01

storage condition

(20% Ethanol)

parameter

217 psi

bed size

6.4 mm × 30 mm

bed volume

1 mL

matrix

polystyrene/divinylbenzene

avg. part. size

15 μm

particle size

15-15 μm

cleaning

1-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

General description

RESOURCE PHE are prepacked SOURCE 15 PHE (phenyl) columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC). For fast, high-resolution purifications, RESOURCE columns are excellent choice for polishing and removal of trace impurities.

Features and Benefits

  • High performance at Low back pressure, even with high sample load of up to 25 mg of Protein.
  • Hydrophobic ligand (phenyl) coupled to the 15 μm monodispersed rigid polystyrene/divinylbenzene matrix.
  • Designed for low- to medium- pressure systems ranging from a peristalic pump to ÄKTA® design and FPLC Systems.
  • RESOURCE HIC columns are available with ether, isopropyl, or phenyl. Generally, RESOURCE PHE will have the strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

FPLC is a trademark of Cytiva
RESOURCE is a trademark of Cytiva
SOURCE is a trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page discusses how to do a purification with SOURCE™ media from Cytiva.

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service