Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0965-10

Phenyl Sepharose 6 Fast Flow / Low Sub

Cytiva 17-0965-10, pack of 25 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€192.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€192.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

€192.00


Please contact Customer Service for Availability

ligand

phenyl

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-0965-10

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14

working range

3-13

suitability

suitable for bioprocess medium

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phenyl Sepharose 6 Fast FLow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification requiring low to medium hydrophobicity.

Application

Phenyl Sepharose 6 Fast FLow is part of the Sepharose Fast FLow HIC platform, which has been an industrial standard for HIC chromatography during recent decades. Phenyl Sepharose 6 Fast FLow is composed of cross-linked 6% Agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages. Two levels of ligand substitution degree help to find the optimal selectivity and binding capacity for a given application.

Phenyl Sepharose 6 Fast FLow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, Phenyl Sepharose 6 Fast FLow meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Standard aromatic HIC media based on Sepharose Fast FLow base matrix derivatized via uncharged, chemically-stable ether linkages.
  • Designed for initial and intermediate purification requiring Low to medium hydrophobicity
  • Two levels of substitution help find optimal selectivity and binding capacity for a given application
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • The hydrophilic nature of the base matrix ensures Low levels of non-specific binding.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimize salt conditions for protein binding and impurity passage in chromatography to achieve required selectivity.

This page discusses column packing and preparation techniques for reverse phase chromatography.

This page shows Hydrophobic Interaction Chromatography (HIC) in a purification strategy.

Column, media, and sample preparation for Hydrophobic Interaction Chromatography (HIC) detailed with Cytiva media.

Protocols

Selectivity of HIC medium from Cytiva is based on properties such as hydrophobicity, porosity and particle size.

Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.

This page discusses various aspects of sample preparation for chromatographic purification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service