Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0855-02

NICK-Column 50 ST

Cytiva 17-0855-02

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€531.00

€531.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€531.00

About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

€531.00


Please contact Customer Service for Availability

packaging

pkg of 50 columns

manufacturer/tradename

Cytiva 17-0855-02

bed size

9 mm × 20 mm

column I.D.

9 mm

particle size

50-150 μm (dry)

capacity

100 μg binding capacity

Related Categories

General description

NICK Columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labeled probe from unincorporated labeled nucleotides.

NICK Columns were originally designed for purification of nick-translated DNA fragments but are also suitable for separation of any labeled probe from unincorporated labeled nucleotides. They are ready for immediate use, and operate by gravity flow. Tests show that at least 97% of radioactivity applied in a nick-translated mixture elutes in two well-separated peaks, corresponding to labeled DNA and unincorporated nucleotides, respectively. At least 90% of the applied DNA is recovered. Maximum sample volume is 100 μ.

Features and Benefits

  • For rapid purification of labeled DNA (20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography.
  • Prepacked with Sephadex® G-50 DNA Grade (see illustra Sephadex® G-50 and G-100 DNA Grade) in distilled water with 0.15% Kathon CG/ICP Biocide.
  • Can accommodate sample volumes up to 100 μ.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sephadex is a registered trademark of Cytiva
illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service