Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0500-01

DEAE-Sephacel

Cytiva 17-0500-01, pack of 500 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€507.00

€507.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€507.00

About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

€507.00


Please contact Customer Service for Availability

description

Ion Exchanger Type (value)

feature

autoclavable In 0.1 M NaCl at 121°C for 30 min

packaging

pack of 500 mL

manufacturer/tradename

Cytiva 17-0500-01

avg. part. size

100 μm

particle size

40-160 μm

cleaning in place

2-12

working range

2-12

capacity

10 mg binding capacity(10 mg Thyroglobulin (MW 669 000)/ml drained medium, 160 mg HSA (MW 68 000)/ml drained medium)

General description

DEAE Sephacel is a weak anion exchanger based on beaded cellulose. DEAE Sephacel is part of the Cytiva range of resins.BioProcess™ chromatography resins are developed andsupported for production scale chromatography. BioProcessresins are produced with validated methods and are tested tomeet manufacturing requirements.

Features and Benefits

  • DEAE Sephacel is supplied preswollen in 20% ethanol.
  • Prepare a slurry by decanting the 20% ethanol solution andreplace it with starting buffer in a ratio of 75% settled resin to25% buffer.
  • The starting buffer must not contain agents whichsignificantly increase the viscosity.
  • The column can beequilibrated with viscous buffers at reduced flow rates afterpacking is completed.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 8 °C. (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service