Skip to Content
Merck
All Photos(1)

Documents

GE17-0471-01

Broad pl Calibration Kit pH 3-10

Cytiva 17-0471-01

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NB.22

form

solid

feature

wetted part: no

packaging

pkg of 10 vials

manufacturer/tradename

Cytiva 17-0471-01

color

White

storage temp.

2-8°C

General description

pl markers are lyophilized mixtures of stable, salt-free, highly purified proteins. The markers may be run in parallel with the experimental samples on isoelectric focusing gels. These markers are not prestained.

The precisely defined pI of the markers allows a permanent and accurate calibration of sample components. pH gradients and pI can be measured to within 0.01 to 0.05 pH unit. The progress of the focusing experiment can be monitored visually by the focusing of methyl red or cytochrome c.

Application

To be used for isoelectric focusing (IEF)

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service