GE17-0420-01
Thiopropyl Sepharose™ 6B
Cytiva 17-0420-01, pack of 15 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
13111000
NACRES:
NA.56
Recommended Products
packaging
pack of 15 g
manufacturer/tradename
Cytiva 17-0420-01
matrix
6% agarose
particle size
45-165 μm
average diameter
90 μm
cleaning in place
2-8
working range
2-8
storage temp.
2-8°C
General description
Thiopropyl Sepharose™ 6B is a medium used for reversible immobilization of molecules containing thiol groups under mild conditions. Optimized for immobilization of small biomolecules.
Application
Convenient immobilization of ligands using pre-activated medium.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sepharose is a trademark of Cytiva
Storage Class Code
"" - ""
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Oksana A Sergeeva et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(4), 1279-1288 (2019-01-06)
The protein acyl transferase ZDHHC5 was recently proposed to regulate trafficking in the endocytic pathway. Therefore, we explored the function of this enzyme in controlling the action of bacterial toxins. We found that ZDHHC5 activity is required for two very
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service