Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0300-05

Ficoll® PM400

Cytiva 17-0300-05, pack of 5 kg

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€7,190.00

€7,190.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€7,190.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

€7,190.00


Please contact Customer Service for Availability

form

solid
spray-dried

packaging

pack of 5 kg

manufacturer/tradename

Cytiva 17-0300-05

color

White

solubility

soluble

General description

High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.

Ficoll® PM400 is a synthetic neutral, highly-branched hydrophilic polymer of sucrose with an average molecular weight of 400 000. It has long been used to form density gradients for separating and isolating eukaryotic cells, organelles and bacterial cells, as a stabilizing agent, and as a preparation medium for isolating mononuclear cells. Applications can also be found in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies.
It is delivered as a spray-dried powder and is therefore readily soluble in aqueous media when it is added slowly with concomitant stirring. Any lumps formed during this procedure will dissolve fairly quickly. Concentrations up to 50% (w/v) can easily be attained

Application

Used in the formation of density gradients for the separation and isolation of eukaryotic cells, organelles, and bacterial cells, as a stabilizing agent and as a separation medium for the isolation of lymphocytes.
It also has applications in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies.
Used for gradient centrifugation in all types of centrifuge rotors and for separation at unit gravity.

Features and Benefits

  • For producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity.
  • Neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution.
  • Concentrations of up to 50% (w/v) covering densities of up to 1.2 g/mL can be achieved.
  • Better osmotic properties than sucrose.
  • Preserves functional and morphological integrity.
  • Useful for separating cells that are sensitive to centrifugation and for separating cells of similar density but different size (under conditions of sedimentation at unit gravity).
  • Does not penetrate biological membranes.
  • Serves as the raw material for preparation of Ficoll-Paque gradients.
  • Used in other applications such as electrophoresis, hybridization, cryopreservation, and as a hapten carrier.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

This page shows methods to remove Percoll from a sample after centrifugation.

The effects of the mononuclear cell separation method described by Bøyum procedure are noted below, since research situations may arise in which they are significant.

Protocols

The following procedure has been evaluated with Ficoll-Paque PLUS and is recommended for separation of normal blood samples for maximum reproducibility.

To prepare a Percoll gradient, the osmolality of Percoll must be adjusted with saline or cell culture medium to make Percoll isotonic with physiological salt solutions.

Related Content

Data was compiled to assist the researcher in selecting references most likely to contain relevant information regarding use of Percoll for a particular cell or tissue type.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service