Skip to Content
Merck
All Photos(1)

Documents

GE17-0220-01

CM-Sephadex® C-50

Cytiva 17-0220-01, pack of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

carboxymethyl

description

Ion Exchanger Type (value)

feature

autoclavable Swollen, in salt form for 30 min at 120°C

packaging

pack of 100 g

manufacturer/tradename

Cytiva 17-0220-01

storage condition

dry media

matrix

cross-linked dextran

particle size

40-120 μm (dry)

cleaning in place

2-13

working range

2-12

capacity

binding capacity

suitability

none detected for bioprocess medium

General description

CM Sephadex® A-50 is a weak cation exchanger based on the well documented and well proven Sephadex® base matrix.
lgG (Mr 160 000)- 7 mg/ml drained medium), Bovine COHb (Mr 69 000)- 140 mg/ml drained medium), Ribonuclease (Mr 13 700)- 120 mg/ml drained medium)

Application

Sephadex® ion exchangers are produced by introducing functional groups onto the cross-linked Dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages.

Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.

Features and Benefits

  • Weak cation exchanger suitable batch techniques.
  • High binding capacities.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service