Skip to Content
Merck
All Photos(2)

Key Documents

GBL204969

Grace Bio-Labs ProPlate MP microtiter plate superstructure

size 96 wells, black polystyrene, adhesive

Synonym(s):

high throughput robotic processing of microarrays, microarray system, microwell plate with permanently affixed well format

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€66.70

€66.70


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€66.70

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black
black polystyrene
round wells
size:
96 wells

€66.70


Please contact Customer Service for Availability

material

black
black polystyrene
round wells

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Grace Bio-Labs 204969

size

96 wells

color

none

Looking for similar products? Visit Product Comparison Guide

General description

N/A

Legal Information

ProPlate MP is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service